Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP2BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557758 Supplier Invitrogen™ Supplier No. PA557758
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNLVEYYYKM KFFTKCVAFS KRIADYDEVH DIPMIAQVTD CLPEFIGRGM AMASFYHARC LQLGLGITRD ETTAKHYYSK ACRLNPALAD ELHSL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.

May act as an adapter that regulates LRP2 function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP2BP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LRP2BP
Gene Accession No. Q9P2M1
Gene Alias 1700113N17Rik; 2310006J04Rik; 4930479L12Rik; KIAA1325; low density lipoprotein receptor-related protein 2 binding protein; Lrp2 binding protein; LRP2-binding protein; Lrp2bp; Megalin-binding protein; megBP; RGD1305896; zgc:165631
Gene Symbols LRP2BP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LRP2BP. Recombinant protein control fragment (Product #RP-96631).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55805
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.