Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 365400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-540-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-540-0 Supplier Invitrogen™ Supplier No. 365400
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

LRP5 Polyclonal Antibody for Western Blot, ICC/IF

LDL related proteins, LRP 5 and LRP 6, are members of the LDL receptor superfamily, which were found to be involved in the canonical Wnt signaling pathway. Mammalian LRP 6 was shown to bind to Wnt 1 and enhance Wnt-induced developmental processes in Xenopus embryos. Mice lacking LRP 6 exhibited developmental defects that are similar to those caused by deficiencies in various Wnt proteins. Recent work also shows that Wnt proteins induce the binding of LRP 5 to Axin, leading Axin degradation and beta catenin stabilization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene LRP5
Gene Accession No. O75197, Q91VN0
Gene Alias BMND1; EVR1; EVR4; HBM; HGNC:6697; LDL receptor related protein 5; low density lipoprotein receptor-related protein 5; low density lipoprotein receptor-related protein 7; low-density lipoprotein receptor-related protein 5; Low-density lipoprotein receptor-related protein 7; LR3; Lrp5; LRP-5; LRP7; LRP-7; mKIAA4142; OPPG; OPS; OPTA1; VBCH2
Gene Symbols LRP5
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the human LRP5 protein. This intracellular region is identical to the corresponding mouse and rat LRP5 amino acid sequences.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16973, 4041
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.