Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-155 Supplier Invitrogen™ Supplier No. PA116913
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-16913 is specific for the ~106 kDa (membrane form) ApoER2 protein. A larger ~130 kDa band may also be seen (glycosylated form). Suggested positive control: mouse brain membrane preparation. Immunogen sequence homology: bovine (95%).

Apolipoprotein E receptor 2 (ApoER2) is an NMDA associated protein found in neurons. It participates in important functions from guiding neurons in the embryonic brain to regulating synaptic signaling. It is also involved in debilitating neurodegenerative diseases such as schizophrenia and Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene LRP8
Gene Accession No. Q14114, Q924X6, Q98931
Gene Alias 4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B
Gene Symbols LRP8
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 863-963).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16975, 396102, 7804
Target Species Human, Mouse, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.