Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598646
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598646 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598646 Supplier Invitrogen™ Supplier No. PA598646
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LRRC20 Polyclonal Antibody for Western Blot, IHC (P), ELISA

LRRC20 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC20
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LRRC20
Gene Accession No. Q8CI70, Q8TCA0
Gene Alias BC036304; leucine rich repeat containing 20; leucine-rich repeat-containing protein 20; lrc20; Lrrc20; RGD1566140; UNQ2429/PRO4989
Gene Symbols LRRC20
Host Species Rabbit
Immunogen Recombinant Human Leucine-rich repeat-containing protein 20 protein (1-184AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216011, 55222
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.