Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRC24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569343
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569343 Supplier Invitrogen™ Supplier No. PA569343
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 85%; Dog: 85%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

LRRC24 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC24
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LRRC24
Gene Accession No. Q50LG9
Gene Alias 6430402H13Rik; leucine rich repeat containing 24; leucine-rich repeat-containing protein 14; leucine-rich repeat-containing protein 24; LRRC14; LRRC14OS; Lrrc24; RGD1308720
Gene Symbols LRRC24
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human LRRC24.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441381
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.