Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC3 Antibody (532820) [Janelia Fluor™ 646], Novus Biologicals™
SDP

Catalog No. FAB5039J
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5039J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB5039J Supplier R&D Systems Supplier No. FAB5039J

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

LRRC3 Monoclonal antibody specifically detects LRRC3 in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen LRRC3
Applications Immunohistochemistry
Classification Monoclonal
Clone 532820
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias C21orf102, chromosome 21 open reading frame 102, leucine rich repeat containing 3, leucine-rich repeat-containing protein 3, LRRC3A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LRRC3, Cys33-Asp204, Accession # Q9BY71
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81543
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.