Learn More
Invitrogen™ LRRC33 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | LRRC33 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | NRROS |
| Gene Accession No. | Q86YC3 |
| Gene Alias | E430025L02Rik; ELLP3030; Garpl1; leucine rich repeat containing 33; leucine-rich repeat-containing protein 33; Leucine-rich repeat-containing protein 33^; Lrcc33; Lrrc33; negative regulator of reactive oxygen species; Nrros; RGD1310771; Transforming growth factor beta activator LRRC33; UNQ3030; UNQ3030/PRO9833 |
| Gene Symbols | NRROS |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.