Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRFIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578282 Supplier Invitrogen™ Supplier No. PA578282
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, NCI-H929 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Leucine-rich repeat FLI-I interacting protein 1 (LRRFIP1/FLAP-1) is a MyD88-interacting signal protein which mediates either induction or repression of TLR signaling. Flap-1 directly interacts with β-catenin and activates β-catenin-dependent transcription activity. Flap-1 or LRRFIP1 was also shown to bind the TNF- -308 promoter polymorphism site, suggesting its possible role in the regulation of TNF- production. FLAP-1 along with LRRFIP2 and FLIIH serve as a balance in coordinating inflammatory response after infection, and may act as a novel target in the control of agonist-induced TLR-mediated inflammatory and autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRFIP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene LRRFIP1
Gene Accession No. Q32MZ4
Gene Alias AU024550; Flap; FLAP (FLI LRR associated protein); FLAP1; flap-1; Fliiap1; FLI-LRR-associated protein 1; GC-binding factor 2; GCF2; GCF-2; H186 FLAP; HUFI1; HUFI-1; leucine rich repeat (in FLII) interacting protein 1; leucine-rich repeat flightless-interacting protein 1; LRR binding FLII interacting protein 1; LRR FLII-interacting protein 1; Lrrfip1; NEDD8-conjugating enzyme; TAR RNA-interacting protein; TRIP
Gene Symbols LRRFIP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human LRRFIP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9208
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.