Learn More
Selleck Chemical LLC LRRK2-IN-1-10mg
SDP
Supplier:  Selleck Chemical LLC S758410MG
Encompass_Preferred
LRRK2-IN-1 is a potent and selective LRRK2 inhibitor with IC50 of 6 nM and 13 nM for LRRK2 (G2019S) and LRRK2 (WT), respectively.
Catalog No. 50-238-9584
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.