Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRRK2 (WT) Recombinant Protein

Catalog No. A15197
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15197 10 μg
A15198 100 μg
A15199 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A15197 Supplier Invitrogen™ Supplier No. A15197
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This full-length, purified and active wild-type LRRK2 protein may be useful in research to better understand the native protein, unaffected by mutations, as well as to explore the mechanisms underlying the disease. Increase disease relevance by interrogating a purified and active full-length LRRK2 protein Uncover novel compounds or hits specific to the full-length form Advance Parkinson's research using the full-length protein and validated assays Related Links:LRRK2 Tools for Advancing Parkinson's Disease ResearchLanthaScreen™ Kinase Activity AssayLanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant, Full Length.

LRRK2 (Leucine Rich Repeat Kinase 2) is a Protein Coding gene. Diseases associated with LRRK2 include Parkinson Disease 8, Autosomal Dominant and Hereditary Late-Onset Parkinson Disease. Among its related pathways are Signaling by PTK6 and ncRNAs involved in Wnt signaling in hepatocellular carcinoma. Gene Ontology (GO) annotations related to LRRK2 (G2019S Mutant) include protein homodimerization activity and transferase activity, transferring phosphorus-containing groups. An important paralog of LRRK2 (G2019S Mutant) is TNNI3K. Mutations in the leucine-rich repeat kinase 2 (LRRK2) gene, which codes for a multidomain protein kinase, are considered the most common genetic cause of late-onset autosomal-dominant Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5S007
Concentration See Label
For Use With (Application) Kinase Assay
Formulation Proprietary buffer with no preservative
Gene ID (Entrez) 120892
Molecular Weight (g/mol) 288 kDa
Name LRRK2 (WT)
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias LRRK2
Common Name LRRK2 (WT)
Gene Symbol LRRK2
Kinase Family LRRK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag DYKDDDDK-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Research Category Neurobiology
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.