Learn More
Invitrogen™ LRRK2 (WT) Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This full-length, purified and active wild-type LRRK2 protein may be useful in research to better understand the native protein, unaffected by mutations, as well as to explore the mechanisms underlying the disease. Increase disease relevance by interrogating a purified and active full-length LRRK2 protein Uncover novel compounds or hits specific to the full-length form Advance Parkinson's research using the full-length protein and validated assays Related Links:LRRK2 Tools for Advancing Parkinson's Disease ResearchLanthaScreen™ Kinase Activity AssayLanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant, Full Length.
Specifications
Specifications
| Accession Number | Q5S007 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | Proprietary buffer with no preservative |
| Gene ID (Entrez) | 120892 |
| Molecular Weight (g/mol) | 288 kDa |
| Name | LRRK2 (WT) |
| Purification Method | Purified |
| Quantity | 100 μg |
| Storage Requirements | -80°C, Avoid Freeze/Thaw Cycles |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.