Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LRSAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121246 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121246 Supplier Invitrogen™ Supplier No. PA5121246
  • $566.00
    $485.65 / Each of 1

    Save $80.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, HT-1080, Mouse brain, Rat spinal cord, Rat brain. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MPLFFRKRKP SEEARKRLEY QMCLAKEAGA DDILDISKCE LSEIPFGAFA TCKVLQKKVL IVHTNHLTSL LPKSCSLLSL ATIKVLDLHD NQLTALPDDL GQLTALQVLN VERNQLMQLP RSIGNLTQLQ TLNVKDNKLK ELPDTVGELR SLRTLNISGN EIQRLPQMLA HVRTLEMLSL DASAMVYPPR EVCGAGTAAI LQFLCKESGL EYYPPSQYLL PILEQDGIEN SRDSPDGPTD RFSREELEWQ NRFSDYEKRK EQKMLEKLEF ERRLELGQRE HTQLLQQSSS.

LRSAM1 is a multifunctional RING finger protein that selectively regulates cell adhesion molecules, has ubiquitin ligase activity, and plays a role in receptor endocytosis and viral budding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRSAM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Lrsam1
Gene Accession No. Q6UWE0, Q80ZI6
Gene Alias CMT2P; E3 ubiquitin-protein ligase LRSAM1; hTAL; leucine rich repeat and sterile alpha motif containing 1; leucine-rich repeat and sterile alpha motif-containing protein 1; LRSAM1; RGD1564403; RIFLE; RING-type E3 ubiquitin transferase LRSAM1; TAL; Tsg101-associated ligase; UNQ6496/PRO21356
Gene Symbols Lrsam1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human LRSAM1 (NP_612370.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227738, 311866, 90678
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.