Learn More
Beckman Coulter LS 13 320 Auto-Prep Station Sample Tubes with Barcode
Designed for use with the LS 13 320 Particle Size Analyzer ALM Auto-Prep Station, which can hold up to 30 sample tubes. The barcode allows programming of the SOM to allow each sample to be automatically run with a designated SOM.
Supplier: Beckman Coulter 8309790
Description
- For use with the AutoPrep Station of the ALM module of the LS 13 320
- Bar coded for easy identification of samples
- Sufficent to fill the AutoPrep Station
- Capacity: 13 mL
Specifications
Sample Tube | |
13 mL | |
30 Pk. | |
LS 13 320 Particle Size Analyzer and Auto-Prep Station |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.