Learn More
Beckman Coulter LS 13 320 XR Single Wavelength (SW) Laser Diffraction Particle Size Analyzer
Single Wavelength (SW) Laser Diffraction Particle Size Analyzer provides expanded measurement range from 0.40 μm to 3,500 μm. The DPS is capable of measuring dry powder samples, while the ULM analyzes samples suspended in aqueous or non-aqueous dilutents.
Supplier:  Beckman Coulter C53023
 View more versions of this product
Catalog No. 15-655-164
Only null left
Add to Cart
Request a Quote
Description
Description
  • Offers particle size distribution data from advanced Polarization Intensity Differential Scattering (PIDS) technology, which enables high-resolution measurements and an expanded dynamic range
  • The XR particle size analyzer provides fast, accurate results, and helps streamline workflows for optimal efficiency
  • Dry Powder System (DPS) Module delivers reliable results from 0.40 μm to 3,500 μm for all types of homogeneous and heterogeneous samples that can be dispersed dry
  • Universal Liquid Module (ULM) analyzes samples suspended in aqueous or non-aqueous dilutents ranging in size from 0.010 μm to 2000 μm
  • Enhanced software simplifies method creation for standardized measurements
  • Provides accurate, reliable detection of multiple particle sizes in a single sample
  • New control standards adequately verify instrument/module performance
  • ADAPT Software features pass/fail results for faster quality control
  • High-security software configuration supports 21 CFR Part 11
  • Direct measurement range from 0.40 μm to 3,500 μm
  • Requires the LS 13 320 XR Workstation or customer-supplied Windows 10 PC that meets Beckman minimum specifications
Specifications
Specifications
0cf2ac13dad753126c4d42dbf0ea921e79eb9079
0.40 μm to 3,500 μm
Requires Controller, Standard LS/HiCap Vacuum, and Optional Sonicator Unit
LS 13 320 XR SW
Particle Size Analysis of Dry Powder, Aqueous, and Non-Aqueous Samples
Universal Liquid Module (ULM) and Dry Powder System (DPS)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.