Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Liquid Scintillation Glass Vials
SDP

Catalog No. 0334025R Shop All DWK Life Sciences Products
Change view
Click to view available options
Material (Liner):
Foamed Polyethylene
Metal Foil
Poly Seal Cone
Closure Material:
Polypropylene
Urea
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Liner) Closure Material
03-340-25R Metal Foil Polypropylene
03-341-25S Poly Seal Cone Urea
03-340-25Q Foamed Polyethylene Polypropylene
03-341-25B Metal Foil Urea
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-340-25R Supplier DWK Life Sciences Supplier No. 986581
Add to Cart
Edge
Add to Cart

Maintain the integrity of seals with a variety of cap liners. Wheaton™ Liquid Scintillation Glass Vials are manufactured from borosilicate glass.

  • Foamed polyethylene liners resilient to acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • Metal foil/cork liners resilient to hydrocarbons, oils, ketones and alcohols
  • Polyethylene cone liners are crack resistant and offer superior torque retention and excellent sealing characteristics
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Metal Foil
Color Clear
Diameter (Metric) Outer 28 mm
Material Borosilicate Glass
Volume (Metric) 20 mL
Closure Material Polypropylene
Quantity 500 Pk.
Closure Size 22-400
Height (Metric) 61 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.