Learn More
MARSHALL SCIENTIFIC Polyscience LS51M11A110C LS5 Benchtop Chiller
SDP
Supplier:  MARSHALL SCIENTIFIC LS51M11A110C
Encompass_Preferred
The LS-Series Compact Chiller provides cooling power for demanding applications and serves as an economical alternative to tap water cooling systems. It features a microprocessor-based controller, digital temperature display (C or °F), and simple operation and maintenance. To optimize cooling efficiency and performance, this sophisticated Chiller also features a modulated refrigeration system. As a result, temperature stability is greatly enhanced and compressor life extended. The Chiller is equipped with either a magnetic drive centrifugal pump or a turbine pump. Wetted parts within the recirculation system are brass, copper, stainless steel, EPDM rubber, Alcryn, nylon, PVC, and polyethylene.
Catalog No. NC2591254
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.