Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM11 Monoclonal Antibody (OTI1A4), TrueMAB™, OriGene
SDP

Catalog No. 501683141
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3141 Supplier OriGene Supplier No. TA809991
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LSM11 is a component of the U7 snRNP complex that is involved in the histone 3′-end pre-mRNA processing. Increases U7 snRNA levels but not histone 3′-end pre-mRNA processing activity, when overexpressed. Required for cell cycle progression from G1 to S phases. Binds specifically to the Sm-binding site of U7 snRNA.

Specifications

Antigen LSM11
Applications Western Blot
Classification Monoclonal
Clone OTI1A4
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LSM11
Gene Accession No. P83369
Gene Alias LSM11, U7 small nuclear RNA associated
Gene Symbols LSM11
Host Species Mouse
Immunogen Full length human recombit protein of human LSM11 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134353
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.