Learn More
NABERTHERM INC LT5/11/B510 TMAX1100C240V
Supplier:  NABERTHERM INC L055H1ANL
Encompass_Preferred
The muffle furnaces are for daily lab use, outstanding workmanship, high level of reliability, non-classified fiber insulation, housing made of structured stainless steel, heating from two sides (three sides L24/11-LT40/12), either flap (L) or lift door (LT)
Catalog No. NC2068351
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.