Learn More
Thermco Products LT5GEO Cellular Data Logger
Catalog No. 04500363
Change view
Click to view available options
Software Type:
Web, Android, Apple IOS
Catalog No. Software Type
04-500-363 Web, Android, Apple IOS
1 options
Catalog No. 04-500-363 Supplier Thermco Products Supplier No. LT5GEO
Add to Cart
Add to Cart
The LT5GEO cellular data logger is for the monitoring of location, humidity, shock, light, and temperature during shipping so you can act immediately.
  • Instant alerts: Get immediate notifications of parameter deviations so you can act immediately to protect your shipment
  • Condition visibility: Know the location and condition of your shipment while it is in transit with location, humidity, shock, light, and temperature sensors
  • Compliance: Meets FCC, CE, DO-160H, and FDA 21 CFR Part 11 standards
Specifications
Type Single-use Datalogger
Display Type Status Indicator Lights
Alarm Types E-mail, Mobile Device, SMS, Audible
Software Type Web, Android, Apple IOS