Learn More
Thermco Products LT5GEO-R Multi-use Cellular Data Logger
Catalog No. 04500364
Change view
Click to view available options
Software Type:
Web, Android, Apple IOS
Catalog No. Software Type
04-500-364 Web, Android, Apple IOS
1 options
Catalog No. 04-500-364 Supplier Thermco Products Supplier No. LT5GEOR
Add to Cart
Add to Cart
The LT5GEO-R multi-use cellular data logger is for the monitoring of location, humidity, shock, light, and temperature during shipping so you can act immediately.
  • Instant alerts: Get immediate notifications of parameter deviations so you can act immediately to protect your shipment
  • Condition visibility: Know the location and condition of your shipment while it is in transit with location, humidity, shock, light, and temperature sensors
  • Compliance: Meets FCC, CE, DO-160H, and FDA 21 CFR Part 11 standards
Specifications
Type Multi-parameter Datalogger
Display Type Status Indicator Lights
Alarm Types E-mail, Mobile Device, SMS, Audible
Software Type Web, Android, Apple IOS