Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LTBR Monoclonal Antibody (5G11)
GREENER_CHOICE

Catalog No. FERMA549059 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549059 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549059 Supplier Invitrogen™ Supplier No. MA549059
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibody 5G11 directed against the mouse LTbetaR is able to activate the LTbetaR in an agonistic way and induces NFkappaB activation and secretion of MIP-2 and IL-6 in mouse fibroblast cells. Therefore, the monoclonal antibody 5G11 can be used to gain more insight into the expression pattern of the LTbetaR and also to investigate molecular mechanisms induced by LTbetaR activation.

The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LTBR
Applications Flow Cytometry, Functional Assay, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 5G11
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene Ltbr
Gene Accession No. P50284
Gene Alias AI256028; CD18; D12S370; LT beta-R; Ltar; LT-beta receptor; LTbetaR; LT-BETA-R; Ltbr; lymphotoxin B receptor; lymphotoxin beta receptor; lymphotoxin beta receptor (TNFR superfamily, member 3); lymphotoxin-beta receptor; TNF receptor-related protein; Tnfbr; TNFCR; TNFR2-RP; TNFR3; TNF-RIII; TNF-R-III; TNFR-III; TNFRrp; TNFR-RP; Tnfrsf3; Tumor necrosis factor C receptor; tumor necrosis factor receptor 2-related protein; Tumor necrosis factor receptor superfamily member 3; tumor necrosis factor receptor superfamily, member 3; tumor necrosis factor receptor type III
Gene Symbols Ltbr
Host Species Rat
Immunogen Drosophila S2 expressed mouse LTbR Ig fusion protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17000
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.