Learn More
Sigma Organic Chemistry L-Tryptophan ethyl ester hydrochloride | 5G | 2899-28-7 | MFCD00038993
Supplier: Sigma Organic Chemistry 936905G

L-Tryptophan ethyl ester hydrochloride, 5G
About This Item:
Linear Formula: C13H16N2O2 -+ HCl
Storage: room temp
EINECS Number: 220-786-0
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.