Learn More
TARGETMOL CHEMICALS INC LTX-315 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6881
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. LTX-315 (Oncopore) the oncolytic peptide inhibits cancer cells through Bax/Bak-regulated mitochondrial membrane permeabilization. purity: 99%
Catalog No. 50-257-6426
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.