Learn More
Bioassay Systems SuperLight™ Luciferase Reporter Gene Assay Kit (500 tests). Bright bioluminescent reagent system for rapid quantitation of luciferase reporter gene expression in transfected cells and high-throughput drug screens.
SDP
Catalog No. 50489305 Shop All Bioassay Systems Products
Encompass_Preferred
Bright bioluminescent reagent system for rapid quantitation of luciferase reporter gene expression in transfected cells and high-throughput drug screens. Key Features: High sensitivity and wide detection range. The emitted light is linear over seven orders of magnitude. Compatible with routine laboratory and HTS formats. Assays can be performed in tubes or microplates, on LJL Analyst, Berthold Luminometer, Top-Count, MicroBeta counters, chemiluminescent image plate readers (CLIPR/LeadSeeker). Assay reagents compatible with all liquid handling systems. Fast and convenient. Homogeneous "mix-and-measure". The optimally combined reagent system allows a single addition step, and simultaneous cell lysis and detection. Robust and amenable to HTS. Z factors of 0.6 to 0.8 are observed in 96-well and 384-well plates. Method: Luminescence. Samples: Cells etc. Species: All. Procedure: Assay takes 2 min. Kit size: 1000 tests. Detection limit: 2 fg luciferase.
Catalog No. 50-489-305 Supplier Bioassay Systems Supplier No. SLLU500
Bright bioluminescent reagent system for rapid quantitation of luciferase reporter gene expression in transfected cells and high-throughput drug screens. Key Features: High sensitivity and wide detection range. The emitted light is linear over seven orders of magnitude. Compatible with routine laboratory and HTS formats. Assays can be performed in tubes or microplates, on LJL Analyst, Berthold Luminometer, Top-Count, MicroBeta counters, chemiluminescent image plate readers (CLIPR/LeadSeeker). Assay reagents compatible with all liquid handling systems. Fast and convenient. Homogeneous "mix-and-measure". The optimally combined reagent system allows a single addition step, and simultaneous cell lysis and detection. Robust and amenable to HTS. Z factors of 0.6 to 0.8 are observed in 96-well and 384-well plates. Method: Luminescence. Samples: Cells etc. Species: All. Procedure: Assay takes 2 min. Kit size: 1000 tests. Detection limit: 2 fg luciferase.