Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LUC7L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109927 Supplier Invitrogen™ Supplier No. PA5109927
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRSHQRSRHS SRDRSRERSK RRSSKERFRD QDLASCDRDR SSRDRSPRDR DRKDKKRSYE SANGRSEDRR SSEEREAGEI.

This gene encodes a protein that contains a C2H2-type zinc finger, coiled-coil region and arginine, serine-rich (RS) domain. A similar protein in mouse interacts with sodium channel modifier 1, and the encoded protein may be involved in the recognition of non-consensus splice donor sites in association with the U1 snRNP spliceosomal subunit. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LUC7L2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.00 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LUC7L2
Gene Accession No. Q7TNC4, Q9Y383
Gene Alias 4930471C18Rik; AA522013; AU015269; CGI59; CGI-59; Cgi74; CGI-74; CGI-74 homolog; CGI-74-like SR-rich; LUC7 like 2, pre-mRNA splicing factor; LUC7B2; LUC7L2; LUC7-like 2; LUC7-like 2 (S. cerevisiae); LUC7-like 2 pre-mRNA splicing factor; Putative RNA-binding protein Luc7-like 2
Gene Symbols LUC7L2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192196, 51631
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.