Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lucifer Yellow CH, Thermo Scientific™

Catalog No. AC229980100
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC229980100 10 mg
AC229980250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC229980100 Supplier Thermo Scientific Chemicals Supplier No. 229980100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67769-47-5
Molecular Formula C13H9Li2N5O9S2
Molecular Weight (g/mol) 457.241
InChI Key RPKCZJYDUKVMGF-UHFFFAOYSA-L
PubChem CID 124220108
ChEBI CHEBI:52106
IUPAC Name dilithium;6-amino-2-(hydrazinecarbonylamino)-1,3-dioxobenzo[de]isoquinoline-5,8-disulfonate
SMILES [Li+].[Li+].C1=C(C=C2C3=C1C(=O)N(C(=O)C3=CC(=C2N)S(=O)(=O)[O-])NC(=O)NN)S(=O)(=O)[O-]

Specifications

Fluorescence Max 531 to 539nm (in H2O)
Merck Index 15, 5653
Infrared Spectrum Authentic
Lambda Max 425 to 431nm (in water)
Formula Weight 457.23
Percent Purity ≥90%
Molar Absorption Coefficient 10700 to 12300L/(mole.cm) (in H2O)
UV Spectrometry Complies
Color Orange
Quantity 10 mg
Physical Form Powder
Chemical Name or Material Lucifer Yellow CH
Show More Show Less
EINECSNumber 267-047-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.