Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lucifer Yellow Polyclonal Antibody
GREENER_CHOICE

Catalog No. A5750
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5750 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5750 Supplier Invitrogen™ Supplier No. A5750
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Lucifer Yellow Polyclonal Antibody for ICC/IF, IHC

Even though the cell soma of a lucifer yellow-filled neuron may be brightly stained, its finer processes can sometimes be faint and may fade rapidly or be obscured by the more intensely stained portions of the neuron. By using anti-lucifer yellow dye antibodies in conjunction with standard enzyme-mediated immunohistochemical methods one can develop a more permanent, fade-free signal for light microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lucifer Yellow
Applications Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 5mM sodium azide; pH 7.2
Gene Alias Lucifer Yellow Dye
Host Species Rabbit
Immunogen Lucifer Yellow.
Purification Method IgG fraction
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.