Learn More
OTT Hydromet Lufft - WS800-UMB Smart Weather Sensor
Supplier:  OTT Hydromet 8381.U01
Encompass_Preferred
First and only compact all-in-one weather sensor with detection of lightning strikes from the WS product family of professional intelligent measurement transducers with digital interface for environmental applications. Integrated design with ventilated radiation protection for measuring: Air temperature, relative humidity, precipitation intensity, precipitation type, precipitation quantity, solar radiation, lightning detection, air pressure, wind direction and wind speed. One external temperature sensor is connectable.
Catalog No. NC1919628
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.