Learn More
Volu Sol Lugol's Iodine Concentrate, 8 mL, Volu-Sol™
SDP
Supplier:  Volu Sol VIG271
Encompass_Preferred
Only the Lugol's solution, part of a two-part solution that includes a Lugol's solution and a dilutent.  Can be used in Gram staining methods.  Also used to test for the presence of starch in organic compounds
Catalog No. NC0964632