Learn More
Description
- Contains complex of Iodine-Polyvinylpyrrolidon (PVP)
- No free iodine, which stabilizes the solution and reduces the evaporation of iodine
- Lugol’s solution and crystal violet form stable complex that will not be released by decolorisation
Specifications
Specifications
| Quantity | 1L |
| Storage Requirements | 15° to 25°C |
| Packaging | Plastic bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.