Learn More
Oz Biosciences LULLABY TRANS REAGENT 3ML
SDP
Supplier:  Oz Biosciences LL73000
Encompass
Lullaby transfection reagent:Lullaby is the ideal siRNA transfection reagent for gene silencing. Relying on the TEE-technology, it has been successfully tested on numerous cell lines, reaching up to 90% gene silencing with high reproducibility and a very low toxicity.
Catalog No. NC1686867
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.