Learn More
Medchemexpress LLC Lumacaftor-d4 | 2733561-44-7 | MFCD00143661 | 99.8% | C24H14D4F2N2O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13262S10MG
Encompass_Preferred
Lumacaftor-d4 is the deuterium-labeled analogue of lumacaftor used as a stable-isotope internal standard and tracer for analytical assays. It supports accurate LC-MS/MS quantitation by providing a consistent mass shift for reliable peak identification and improved precision in bioanalytical workflows.
  • Deuterium-labeled stable isotope internal standard
  • Compatible with LC-MS/MS methods
  • High isotopic and chemical purity for reliable quantitation
  • Available in small, pre-weighed quantities for analytical use
  • Suitable as a tracer in pharmacokinetic and bioanalytical studies
Catalog No. 50-002-59690
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.