Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Lumi Rod (Light Pipe)
SDPSCI_ED

Catalog No. S89544 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S89544 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89544 Supplier United Scientific™ Supplier No. LUMRD48
Add to Cart
Add to Cart
This item is not returnable. View return policy

The United Scientific™ Lumi Rod is an excellent tool for demonstrating the principle of fiber optics by internal reflection.

The United Scientific Lumi Rod is an excellent tool for demonstrating the principle of fiber optics by internal reflection.

  • The curled 48 in. long clear acrylic rod conducts light by internal reflection
  • A flashlight or laser pointer can be used for the demonstration

Specifications

Grade 5 to 12
Format Kit
Product Type Light Pipe
Height (English) 48 in.
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.