Learn More
Enzo Life Sciences Luminol (1g). CAS: 521-31-3
SDP
Supplier:  Enzo Life Sciences ALX610002G001
Encompass_Preferred
Key reagent for generating luminescence. A reagent for the highly sensitive and selective detection of nitric oxide (NO) in biological samples by a luminol-H2O2 chemiluminescence method. Alternative name: 5-Amino-2,3-dihydro-1,4-phthalazinedione, 3-Aminophthalhydrazide. Purity: ≥97% (HPLC). Appearance: Off-white to brown solid. Solubility: Soluble in DMSO (50mg/ml) or in 10% ammonia (hot, 100mg/ml). Long Term Storage: Ambient.
Catalog No. 50-200-9592
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.