Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Corporation Lumit HMGB1 (Human/Mouse) Immunoassay

Catalog No. PRW6112 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Detection Method:
Bioluminescence
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
PRW6112 Bioluminescence
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRW6112 Supplier Promega Corporation Supplier No. W6112
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

No-Wash Immunoassay for HMGB1 Detection

HMGB1 functions as a DAMP released from cells in inflammatory responses. Lumit technology simplifies HMGB1 quantitation with an add-mix-measure protocol completed in 90 minutes or less.

Specifications

CE Marker No
Clia Complexity Not Applicable
Detectable Analytes HMGB1
Detection Method Bioluminescence
Format Kit
Sample Type Cell Culture Supernatants
Sensitivity High
Type Immunoassay
For Use With (Application) Research
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.