Learn More
Adam Equipment LPB 223e Luna Precision Balance 220g capacity x 0.001g readability
Supplier:  Adam Equipment LPB 223E
Encompass_Preferred
Adam Equipment BCT Label Printing Scales feature a removable, easy-to-clean stainless steel platform, BCT advanced label printing scales provide excellent value for industrial counting and weighing tasks. The BCT features a printer that can print weight, QR codes, bar codes and labels, which can be customized using the included software. Store and recall up to 1006 PLUs or piece weights to improve efficiency in production or warehouse applications. RS-232 interface provide speedy connection to computers and printers. BCT offers a choice of two platform sizes, making it ideal for counting inventory, shipping and receiving, and batching items for processing. With an easy-to-clean stainless steel platform, BCT scales offer excellent value for industrial counting tasks. BCT contains a printer that prints QR codes, bar codes and labels. Store up to 1006 PLUs or piece weights to expedite inventory and batching jobs. Rechargeable battery included for operation.
Catalog No. NC1992987
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.