Learn More
United Scientific Supplies Lung Apparatus
SDPSCI_ED
Catalog No. S27829 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Activity Guide
Catalog No. Includes
S27829 Activity Guide
1 options
Catalog No. S27829 Supplier United Scientific Supplies Supplier No. LUNG01B
Add to Cart
Add to Cart
Demonstrates how lungs are filled and emptied with air through contraction and relaxation of the diaphragm muscle
  • Demonstrates how contraction and relaxation of the diaphragm muscle (rubber sheet) fills and empties lungs (balloons)
  • Activity guide included
Specifications
Includes Activity Guide
Material Rubber Sheet (Diaphragm Muscle), Balloons (Lungs)
Product Type Model, Lung