Learn More
ATCC Lung Cancer Cell Line Panel
Supplier:  ATCC TCP1016
Encompass
A panel of 7 lung cancer cell lines with varying degrees of genetic complexity. They have genomic mutations in one or more of the following genes according to the Sanger COSMIC database: CDKN2A, EGFR, KRAS, NRAS, PIK3CA, PIK3R1, SMARCA4, STK11, and TP53.
Catalog No. NC1829391
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.