Learn More
Apexbio Technology LLC Luotonin A 205989-12-4 1mg
SDP
Supplier:  Apexbio Technology LLC C44331
Encompass_Preferred
Luotonin A (CAS 205989-12-4) is a small-molecule inhibitor targeting human topoisomerase I It is designed to stabilize the topoisomerase I-DNA binary complex thereby interfering with DNA processing pathways Luotonin A exerts its biological activity primarily through inhibition of DNA strand religation promoting the accumulation of topoisomerase I-linked DNA breaks and inducing topoisomerase I-dependent cytotoxicity In in vitro studies Luotonin A demonstrates inhibitory activity with an IC50 value of approximately 1 8 g/mL in P-388 cells Based on these pharmacological properties Luotonin A holds research potential in cancer research to investigate topoisomerase I-mediated pathways and DNA damage response
Catalog No. 50-000-91624
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.