Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Lupeol

Catalog No. AC465500010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465500010 1 g
AC465502500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465500010 Supplier Thermo Scientific Chemicals Supplier No. 465500010
Add to Cart

CAS: 545-47-1 | C30H50O | 426.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545-47-1
Molecular Formula C30H50O
Molecular Weight (g/mol) 426.73
MDL Number MFCD00017351
InChI Key MQYXUWHLBZFQQO-JGGBMTAGSA-N
IUPAC Name (1R,3aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a,5a,5b,8,8,11a-hexamethyl-1-(prop-1-en-2-yl)-icosahydro-1H-cyclopenta[a]chrysen-9-ol
SMILES CC(=C)[C@@H]1CC[C@]2(C)CCC3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12

Specifications

Alpha Vector LUPEOL
Melting Point 206°C to 210°C
Color White
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 1 g
Solubility Information practically insoluble aq. soln., freely soluble in ether, benzene, petroleum ether, warm alcohol, practically insoluble in chloroform, methanol
Formula Weight 426.72
Physical Form Powder
Chemical Name or Material Lupeol
ShelfLife 3 years
storageNote1 Freezer -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.