Learn More
Bioworld Luria-Bertani (LB) Agar, Miller, 20-Gram Capsules, 50 Capsule(s)
Supplier:  Bioworld 306201132
Encompass_Preferred
Luria-Bertani (LB) Agar, MillerFor propagation and maintenance of Escherichia coliConvenient premeasured and pre-mixed capsulesContains twenty times the sodium chloride level of Luria Agar Base, Miller and twice the level found in LB Agar, LennoxpH: 7.0Working Concentration: 40 g/L (2 capsules)Prepared Appearance: light amber agarSoluble in boiling waterContains:TryptoneYeast ExtractSodium ChlorideAgarThis product is sold for laboratory research use only.
Catalog No. 50-254-417
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.