Learn More
Cayman Chemical Lutein, 127-40-2, 1 mg
SDP
Supplier:  Cayman Chemical 100108111
Encompass_Preferred
A natural yellow carotenoid, which can be found in plants, egg yolks, and in the human retina; a lipid-soluble antioxidant that can circulate in the plasma and confer cardioprotective, anti-inflammatory, and anti-angiogenic effects
Catalog No. NC1453928
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.