Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lutetium pieces, 99.9% (REO)

Catalog No. aa0062803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0062803 1 g
AA0062806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0062803 Supplier Thermo Scientific Chemicals Supplier No. 00062803
Only null left
Add to Cart

CAS: 7439-94-3 | Lu | 174.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in air-stable glass bottle
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-94-3
Molecular Formula Lu
Molecular Weight (g/mol) 174.97
MDL Number MFCD00011098
InChI Key OHSVLFRHMCKCQY-UHFFFAOYSA-N
Synonym lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
PubChem CID 23929
ChEBI CHEBI:33382
IUPAC Name lutetium
SMILES [Lu]

Specifications

Quantity 1 g
Odor Odorless
Packaging Glass bottle
Form Pieces
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Lutetium pieces
Hazard Category H228
Precautionary Statement P210-P240-P241-P280-P370+P378q
EINECSNumber 231-103-0
TSCA Yes
storageNote1
  • Packaged in air-stable glass bottle
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.