Learn More
StatLab LUXOL FAST BLUE/ CRESYL VIOLET
SDP
Supplier:  StatLab KTLFBPT
Encompass_Preferred
This kit stains myelin sheaths and nissl substances in one hour and 15 minutes. Stain kit includes Luxol Fast Blue Stain, Cresyl Echt Violet Stain, 70% Reagent Alcohol, 0.05% Lithium Carbonate.
Catalog No. NC0657719
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.