Learn More
StatLab LUXOL FAST BLUE KIT
SDP
Supplier:  StatLab KTLFB
Encompass_Preferred
Luxol Fast Blue-Cresyl Echt Violet Stain Kit, Kit Consists of: Luxol Fast Blue Stain 100mL, Cresyl Echt Violet Stain 100mL, 70% Reagent Alcohol Sol. Pint, Lithium Carbonate Sol, An excellent method for Myelin Sheath and Nissl Substance
Catalog No. NC9030259
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.