Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LXR alpha/beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674650
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-650 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-650 Supplier Thermo Scientific Supplier No. PA19066
  • $588.00
    $487.70 / Each of 1

    Save $100.30

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

LXR alpha/beta Polyclonal Antibody

LXR alpha/beta Polyclonal Antibody for Western Blot, ICC/IF, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen LXR alpha/beta
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NR1H3
Gene Accession No. P55055, Q13133
Gene Alias AI194859; AU018371; liver X nuclear receptor alpha variant 1; liver X nuclear receptor beta; Liver X Receptor; liver X receptor alpha; Liver X receptor beta; LX receptor beta; LXR; LXR alpha; LXR beta; LXRA; LXR-a; LXRalpha; LXRB; LXR-b; LXRbeta; NER; NER1; NER-I; NR1H2; NR1H3; nuclear orphan receptor LXR-alpha; nuclear orphan receptor LXR-beta; nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 2; nuclear receptor subfamily 1, group H, member 3; OR-1; Orphan nuclear receptor OR-1; oxysterols receptor LXR-alpha; Oxysterols receptor LXR-beta; retinoid X receptor-interacting protein No.15; Rip15; RLD1; RLD-1; steroid hormone-nuclear receptor NER; ubiquitous receptor; ubiquitously-expressed nuclear receptor; ubiquitously-expressed nuclear receptor 1; ubiquitously-expressed nuclear receptor 2; Unr; Unr1; Unr2; UR
Gene Symbols NR1H2, NR1H3
Host Species Goat
Immunogen Internal sequence corresponding to amino acids CRLQDKKLPPLLSEI.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10062, 7376
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.