Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LXR alpha Recombinant Rabbit Monoclonal Antibody (12H21L12)

Catalog No. 703430
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-343-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-343-0 Supplier Invitrogen™ Supplier No. 703430
  • $502.00
    $376.65 / Each of 1

    Save $125.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Rat, Bovine.

NR1H3 (oxysterols receptor LXR-alpha) is a nuclear receptor that exhibits a ligand-dependent transcriptional activation activity. Its interaction with retinoic acid receptor (RXR) shifts RXR from its role as a silent DNA-binding partner to an active ligand-binding subunit in mediating retinoid responses through target genes defined by LXRES. LXRES are DR4-type response elements characterized by direct repeats of two similar hexanucleotide half-sites spaced by four nucleotides. NR1H3 plays an important role in the regulation of cholesterol homeostasis, regulating cholesterol uptake through MYLIP-dependent ubiquitination of LDLR, VLDLR, and LRP8. It also interplays functionally with RORA for the regulation of genes involves in liver metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LXR alpha
Applications Western Blot
Classification Recombinant Monoclonal
Clone 12H21L12
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene NR1H3
Gene Accession No. Q13133, Q9Z0Y9
Gene Alias AU018371; liver X nuclear receptor alpha variant 1; liver X receptor alpha; LXR; LXR alpha; Lxra; LXR-a; LXRalpha; LXRb; NER; NERI; NR1H2; Nr1h3; nuclear orphan receptor LXR-alpha; nuclear oxysterol receptor LxR-alpha; nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 3; oxysterols receptor LXR-alpha; RLD1; RLD-1; ubiquitously-expressed nuclear receptor 1; Unr1
Gene Symbols NR1H3
Host Species Rabbit
Immunogen Peptides corresponding to human LXR alpha [1) aa71-aa88 2) aa317-aa330 3) aa177-aa195].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10062, 22259
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.