Learn More
Apexbio Technology LLC LY 2365109 hydrochloride 1779796-27-8 50mg
SDP
Supplier:  Apexbio Technology LLC B739450
Encompass_Preferred
LY 2365109 hydrochloride (CAS 1779796-27-8) is a selective inhibitor of glycine transporter 1 (GlyT1) functioning by specifically blocking GlyT1-mediated glycine uptake This inhibition leads to elevated glycine concentrations in the synaptic cleft thereby modulating NMDA receptor activity In cellular assays LY 2365109 hydrochloride has demonstrated nanomolar potency against human GlyT1 The compound serves as a valuable tool for investigating the physiological role of glycine transport pathways in the central nervous system and for exploring the involvement of GlyT1 in the mechanisms underlying neuropsychiatric disorders
Catalog No. 50-000-89024
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.