Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-49A Monoclonal Antibody (A1 (Ly49A)), PE, eBioscience™
GREENER_CHOICE

Catalog No. 5010909
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-109-09 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-109-09 Supplier Invitrogen™ Supplier No. 12585682
  • $372.00
    $279.65 / Each of 1

    Save $92.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This A1 monoclonal antibody reacts with Ly49A, a 44-kDa member of the Ly49 family of type II transmembrane receptors expressed on natural killer (NK) cells. This receptor is present as a disulfide-linked homodimer with C-type lectin extracellular domains. An inhibitory receptor, Ly49A binds MHC class I molecules, such as H-2Dd, H-2Dk, and H-2DP, to inhibit NK-mediated cytotoxicity by triggering phosphorylation of its ITIM domain and recruitment of SHP-1 and SHP-2. Ly49A is expressed on DX5+CD3- NK cells and DX5+CD3+ NK T cells, as well as on a population of memory CD8+ T cells in C57BL/6, C57BL/10, and B10 congenic mice. Applications Reported: This A1 (Ly49A) antibody has been reported for use in flow cytometric analysis. Applications Tested: This A1 (Ly49A) antibody has been tested by flow cytometric analysis on C57Bl/6 splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Ly49A is a gene that encodes a protein expressed on the surface of natural killer cells, which is involved in recognizing and eliminating virally infected or cancerous cells. It is located on chromosome 6 and is part of a larger family of genes that encode for similar receptors. The Ly49A protein interacts with a variety of ligands, including certain viruses and tumor cells, and is involved in regulating the activation and function of natural killer cells. Mutations or variations in the Ly49A gene have been associated with various diseases and conditions, including autoimmune disorders and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49A
Applications Flow Cytometry
Classification Monoclonal
Clone A1 (Ly49A)
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Klra1
Gene Accession No. P20937
Gene Alias A1; CH29-493D4.3; class I MHC receptor KLRA22; killer cell lectin like receptor A1, pseudogene; killer cell lectin-like receptor 2; killer cell lectin-like receptor, subfamily A, member 1; killer cell lectin-like receptor, subfamily A, member 2; KLRA#; KLRA1; KLRA1P; Klra2; Klra22; KLRAP1; Ly49; Ly49 inhibitory receptor 8; Ly49a; ly-49a; Ly49i8; LY49L; Ly-49L; Ly49o>129>; Ly49v; lymphocyte antigen 49a; natural killer cell receptor Ly49A; novel killer cell lectin-like receptor subfamily A protein; t lymphocyte antigen A1; T-cell surface glycoprotein YE1/48
Gene Symbols Klra1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16627
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.