Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-49E/F Monoclonal Antibody (CM4), APC, eBioscience™
GREENER_CHOICE

Catalog No. 5015225
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-152-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-152-25 Supplier Invitrogen™ Supplier No. 17584880
  • $170.00
    $127.65 / Each of 1

    Save $42.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: This CM4 monoclonal antibody reacts with a subset of mouse Ly49 receptors, specifically Ly49E and Ly49F. The Ly49 family of receptors are clustered in the Natural Killer Cell complex on chromosome 6. Expression is confined to NK cell, lymphocytes that play a key role in protection from infection and malignant diseases as well as graft rejection and autoimmune diseases. Ly49E is unique from other family members in that it contains an ITIM (immunoreceptor tyrosine-based inhibitory motif) domain and is expressed on all mouse strains. Developmentally Ly49E is one of the first markers detected on NK cells but expression decreases after birth to about 1% of NK cells in the adult spleen. Upon stimulation, expression can again be induced on Ly49E negative cells. The CM4 antibody is a useful reagent for the characterization of Ly49E expression when used in combination with other Ly49 antibodies such as 14B11 (Ly49C/I/F/H), 3D10 (Ly49C), YLI-90 (Ly49I), and A1 (Ly49A). Applications Reported: This CM4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CM4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be care...

The Ly49 family of receptors are clustered in the Natural Killer Cell complex on chromosome 6. Expression is confined to NK cell, lymphocytes that play a key role in protection from infection and malignant diseases as well as graft rejection and autoimmune diseases. Ly49E is unique from other family members in that it contains an ITIM (immunoreceptor tyrosine-based inhibitory motif) domain and is expressed on all mouse strains. Developmentally Ly49E is one of the first markers detected on NK cells but expression decreases after birth to about 1% of NK cells in the adult spleen. Upon stimulation, expression can again be induced on Ly49E negative cells. The CM4 antibody is a useful reagent for the characterization of Ly49E expression when used in combination with other Ly49 antibodies such as 14B11 (Ly49C/I/F/H), 3D10 (Ly49C), YLI-90 (Ly49I), and A1 (Ly49A).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49E/F
Applications Flow Cytometry
Classification Monoclonal
Clone CM4
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Klra5
Gene Accession No. Q60652, Q60653
Gene Alias class I MHC receptor KLRA5; killer cell lectin-like receptor 5; Killer cell lectin-like receptor 6; killer cell lectin-like receptor, subfamily A, member 5; killer cell lectin-like receptor, subfamily A, member 6; Klra5; Klra6; Ly49e; Ly-49e; Ly49-e; Ly49E receptor; Ly49f; ly-49f; Ly49-f; Lymphocyte antigen 49e; lymphocyte antigen 49f; T-cell surface glycoprotein Ly-49E; T-cell surface glycoprotein Ly-49F
Gene Symbols Klra5, Klra6
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16636, 16637
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.